Learn More
Abcam BCIP toluidine salt, histochemical alkaline phosphatase substrate, 500MG
Catalog No. 502704824
Encompass_Preferred
MW 433.62 g/mol, Purity >99%. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4824 Supplier Abcam Supplier No. AB146225500MG
MW 433.62 g/mol, Purity >99%. Chromogenic substrate for alkaline phosphatase. Often used in combination with nitro blue tetrazolium chloride (NBT) to amplify the sensitivity of staining.
The product is subject to the following: Abcam Restricted Use Statement