Learn More
Research Products International Corp [X-Phos] [BCIP] 5-Bromo-4-chloro-3-indolyl-phosphate, disodium salt, 1 Gram
SDP
Catalog No. 50213197
Encompass_Preferred
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.
Catalog No. 50-213-197 Supplier Research Products International Corp Supplier No. B740001.0
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.