Learn More
Supply Solutions BCIP, pkg of 3 mL (150 mg)
Catalog No. 501003395
Encompass
BCIP serves as a substrate for alkaline phosphatase. The reaction product has a blue color and is insoluble in water. The blue color can be visually seen.
Catalog No. 50-100-3395 Supplier Supply Solutions Supplier No. SIGMA 11383221001
BCIP serves as a substrate for alkaline phosphatase. The reaction product has a blue color and is insoluble in water. The blue color can be visually seen.