Learn More
Cayman Chemical BCIP (p-toluidine salt), Purity: ≥95%, CAS: 6578-06-9, 500 mg
SDP
Catalog No. 502950615
Encompass_Preferred
A colorimetric substrate for alkaline phosphatase; can be used to detect alkaline phosphatase activity by IHC or WB
Catalog No. 50-295-0615 Supplier Cayman Chemical Supplier No. 34423500 MG
A colorimetric substrate for alkaline phosphatase; can be used to detect alkaline phosphatase activity by IHC or WB