Learn More
New England Biolabs, Inc. BciVI – 1000 units
SDP
Catalog No. 50812313
Encompass
A restriction endonuclease that recognizes the sequence GTATCCNNNNN_N^.
Catalog No. 50-812-313 Supplier New England Biolabs, Inc. Supplier No. R0596L
A restriction endonuclease that recognizes the sequence GTATCCNNNNN_N^.