Learn More
MSC Miller By Honeywell Full Body Harness, Polyester, Quick-Connect Buckles, Size 2XL/3XL, 1 ea
Catalog No. 5000507682
Encompass
This full body harness features breathable, open-core padding technology designed to improve airflow and reduce heat and moisture build-up during use. Constructed with durable polyester webbing, this harness is built for comfort and security in demanding work environments.
  • Features side and back D-rings for versatile attachment
  • Utilizes quick-connect buckles on chest and leg straps for easy donning
  • Includes cam-style shoulder strap buckles
  • Equipped with back, leg, and shoulder padding for enhanced comfort
  • Design minimizes contact around the neck and thighs to reduce chafing
  • Supports a capacity of 420 lbs. with a tensile strength of 5,000 lbs.
  • Meets OSHA 1910.66, OSHA 1926.502(d), ANSI A10.32, and ANSI Z359.11 standards
Catalog No. 50-005-07682 Supplier MSC Supplier No. 80890791
This full body harness features breathable, open-core padding technology designed to improve airflow and reduce heat and moisture build-up during use. Constructed with durable polyester webbing, this harness is built for comfort and security in demanding work environments.
  • Features side and back D-rings for versatile attachment
  • Utilizes quick-connect buckles on chest and leg straps for easy donning
  • Includes cam-style shoulder strap buckles
  • Equipped with back, leg, and shoulder padding for enhanced comfort
  • Design minimizes contact around the neck and thighs to reduce chafing
  • Supports a capacity of 420 lbs. with a tensile strength of 5,000 lbs.
  • Meets OSHA 1910.66, OSHA 1926.502(d), ANSI A10.32, and ANSI Z359.11 standards