Learn More
Aviva Systems Biology Corp BCKDHA Antibody - N-terminal region (ARP56119_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56119_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCKDHA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BCKDHA. Alternate names: FLJ45695, MSU, MSUD1, OVD1A, BCKDE1AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61726