Learn More
Advansta Inc BCKGRND QUENCHING SHEETS (10)
Catalog No. NC1623617
Encompass
Background Quenching Sheets. 10 sheets. An added layer to decrease background. For improved imaging of blots and gels
Catalog No. NC1623617 Supplier Advansta Inc Supplier No. L07001010
Background Quenching Sheets. 10 sheets. An added layer to decrease background. For improved imaging of blots and gels