Learn More
Enzo Life Sciences Bcl-2 monoclonal antibody (83 8B) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM072E
Encompass_Preferred
Clone: 83-8B. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat Bcl-2 β. Species reactivity: Human, Mouse, Rat Drosophila. Applications: Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7134