Learn More
Biocare Medical, LLC Bcl-6 [LN22] Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM410C
Encompass_Preferred
BCL-6 Mouse Monoclonal Antibody is commonly expressed in diffuse large cell lymphomas, follicular lymphomas and Burkitt lymphoma/leukemia. Bcl-6 protein is expressed mainly by follicle center cells, a few interfollicular T lymphocytes and in nodular lymphocyte predominant Hodgkin disease. However Blc-6 mouse monoclonal is not expressed in hairy cell leukemia, mantle cell or marginal-zone derived lymphomas. In humans, Bcl-6 encodes for a Kruppel-type zinc finger protein that is believed to be important in germinal center formation. The LN22 clone of Bcl-6 provides superior sensitivity compared to [PIF6].The Bcl-6 gene, originally cloned from a tumor with 3q27 translocation, is commonly expressed in large cell lymphomas. In humans, Bcl-6 encodes for a Kruppel-type zinc finger protein that is believed to be important in germinal center formation.
Catalog No. 50-826-98