Learn More
Enzo Life Sciences Bcl-w monoclonal antibody (16H12) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804354C100
Encompass_Preferred
Clone: 16H12. Isotype: IgG2a. Immunogen: Recombinant human Bcl-w. Species reactivity: Human, Mouse, Rat, Monkey, Dog. Applications: IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 25% glycerol and 0.02% sodium azide.
Catalog No. 50-200-9879