Learn More
Enzo Life Sciences Bcl-XL monoclonal antibody (1C2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804660C100
Encompass_Preferred
Clone: 1C2. Isotype: IgG2a κ. Immunogen: Recombinant mouse Bcl-XL (aa 1-209). Species reactivity: Human, Mouse. Applications: ELISA, Flow Cytometry, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9967