Learn More
Enzo Life Sciences Bcl xL monoclonal antibody (2H12) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM080E
Encompass_Preferred
Clone: 2H12. Host: Mouse. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human Bcl-xL. UniProt ID: Q07817. Species reactivity: Human, Mouse, Rat. Applications: WB. Recommended Dilutions: Western Blot (1-5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ascites. Formulation: Liquid. In PBS containing 0.08% sodium azide.
Catalog No. 50-200-7135