Learn More
Medchemexpress LLC Bcl-XL Mouse His 50ug
SDP
Supplier:  Medchemexpress LLC HYP7284650UG
Encompass_Preferred
Bcl-XL protein forms heterodimers with BAX BAK or BCL2 but its antiapoptotic activity does not require heterodimerization with BAX Bcl-XL Protein Mouse (His) is the recombinant mouse-derived Bcl-XL protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-81877