Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 8915MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM520. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: WEHI-231 or Ramos cells or lymphoma. Cellular Staining: Nuclear, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human BCL10 recombinant protein (epitope aa122-168). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 33kDa.Chromosome: 1p22.3. Gene Symbol: BCL10. Gene ID: 8915. Ensemble: ENSG00000142867. Uniprot: XP_011540699.
Catalog No. 50-188-5313