Learn More
Aviva Systems Biology Corp BCL11A Antibody - C-terminal region (ARP39569_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL11A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BCL11A. Alternate names: BCL11A-L, BCL11A-S, BCL11A-XL, CTIP1, EVI9, FLJ10173, FLJ34997, HBFQTL5, KIAA1809, ZNF856ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-815