Learn More
Aviva Systems Biology Corp BCL11A Antibody - N-terminal region (ARP39322_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39322_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL11A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BCL11A. Alternate names: BCL11A-L, BCL11A-S, BCL11A-XL, CTIP1, EVI9, FLJ10173, FLJ34997, HBFQTL5, KIAA1809, ZNF856ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-56985