Learn More
Aviva Systems Biology Corp BCL11B Antibody - middle region (ARP33293_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33293_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL11B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BCL11B. Alternate names: CTIP-2, CTIP2, RIT1, hRIT1-alpha, ZNF856BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54949