Learn More
Aviva Systems Biology Corp BCL11B Antibody - C-terminal region (ARP39820_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39820_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL11B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BCL11B. Alternate names: CTIP-2, CTIP2, RIT1, hRIT1-alpha, ZNF856BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-833