Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 596MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: BCL2/782 + BCL2/796. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Jurkat, K562, HL-60, or HeLa Cells. Tonsil or follicular lymphomas. Cellular Staining: Outer mitochondrial membranes, endoplasmic reticulum as well as nuclear membranes. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human bcl-2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 25-26kDa.Chromosome: 18q21.33. Gene Symbol: BCL2. Gene ID: 596. Ensemble: ENSG00000171791. Uniprot: XP_011524437.
Catalog No. 50-188-5135