Learn More
Aviva Systems Biology Corp BCL2 Antibody - N-terminal region (AVARP01005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP01005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human BCL2. Alternate names: Bcl-2, PPP1R50ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62462