Learn More
ATCC BCL2 Jurkat; Acute T Cell Leukemia; Human (Homo sapiens)
Catalog No. 502384228
Encompass
BCL2 Jurkat is a T lymphocyte cell line that was isolated in 2003 from the peripheral blood of a 14-year-old, male patient with leukemia, acute T cell. It was derived by transfecting the human Jurkat T cells with the pSFFV-neo mammalian expression vector containing the human BCL-2 ORF insert and a neomycin-resistant gene. This cell line was deposited by Stanley Korsmeyer and can be used in immunology and immune system disorder research.
Catalog No. 50-238-4228 Supplier ATCC Supplier No. CRL2899
Add to Cart
Add to Cart
BCL2 Jurkat is a T lymphocyte cell line that was isolated in 2003 from the peripheral blood of a 14-year-old, male patient with leukemia, acute T cell. It was derived by transfecting the human Jurkat T cells with the pSFFV-neo mammalian expression vector containing the human BCL-2 ORF insert and a neomycin-resistant gene. This cell line was deposited by Stanley Korsmeyer and can be used in immunology and immune system disorder research.