Learn More
Aviva Systems Biology Corp BCL2A1 Antibody - C-terminal region (AVARP09047_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09047_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL2A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BCL2A1. Alternate names: GRS, BFL1, ACC-1, ACC-2, HBPA1, BCL2L5ApplicationWB. Species reactivity: Dog, Pig, Rat
Catalog No. 50-014-62586