Learn More
Aviva Systems Biology Corp BCL2L1 Antibody - N-terminal region (AVARP02008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCL2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BCL2L1. Alternate names: BCLX, BCL2L, BCLXL, BCLXS, Bcl-X, bcl-xL, bcl-xS, PPP1R52, BCL-XL/SApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit, Sheep
Catalog No. 50-014-62474