Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 604MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: BCL6/1527. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Raji or Ramos cells. Tonsil or Hodgkin s lymphoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human bcl-6 protein fragment (aa256-389). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 95kDa.Chromosome: 3q27.3. Gene Symbol: BCL6. Gene ID: 604. Ensemble: ENSG00000113916. Uniprot: NP_001124317.
Catalog No. 50-188-5148