Learn More
Medchemexpress LLC BCL6-IN-4 | 2253879-65-9 | 99.7% | 503.04 | 1 ML
SDP
Catalog No. 5000352294
Encompass_Preferred
BCL6-IN-4 is a potent B-cell lymphoma 6 (BCL6) inhibitor with an IC50 of 97 nM. It demonstrates anti-tumor activities.
  • Potent B-cell lymphoma 6 (BCL6) inhibitor
  • IC50 of 97 nM against BCL6
  • Demonstrates anti-tumor activities
  • Inhibits OCI-Ly1 and OCI-Ly3 cell lines growth with GI50 values of 2.8 μM and 4.2 μM, respectively
Catalog No. 50-003-52294 Supplier Medchemexpress LLC Supplier No. HY13664010MM/1ML
Add to Cart
Add to Cart
BCL6-IN-4 is a potent B-cell lymphoma 6 (BCL6) inhibitor with an IC50 of 97 nM. It demonstrates anti-tumor activities.
  • Potent B-cell lymphoma 6 (BCL6) inhibitor
  • IC50 of 97 nM against BCL6
  • Demonstrates anti-tumor activities
  • Inhibits OCI-Ly1 and OCI-Ly3 cell lines growth with GI50 values of 2.8 μM and 4.2 μM, respectively