Learn More
Medchemexpress LLC BCL6-IN-4 | 2253879-65-9 | 99.7% | 503.04 | 100 MG
SDP
Catalog No. 5000347963
Encompass_Preferred
BCL6-IN-4 is a potent inhibitor of B-cell lymphoma 6 (BCL6), with an IC50 of 97 nM. This compound demonstrates anti-tumor activities. It effectively inhibits the growth of OCI-Ly1 and OCI-Ly3 cell lines, with GI50 values of 2.8 μM and 4.2 μM respectively.
  • Potent BCL6 inhibitor
  • Exhibits anti-tumor activities
  • Inhibits OCI-Ly1 cell line growth with a GI50 of 2.8 μM
  • Inhibits OCI-Ly3 cell line growth with a GI50 of 4.2 μM
Catalog No. 50-003-47963 Supplier Medchemexpress LLC Supplier No. HY136640100MG
BCL6-IN-4 is a potent inhibitor of B-cell lymphoma 6 (BCL6), with an IC50 of 97 nM. This compound demonstrates anti-tumor activities. It effectively inhibits the growth of OCI-Ly1 and OCI-Ly3 cell lines, with GI50 values of 2.8 μM and 4.2 μM respectively.
  • Potent BCL6 inhibitor
  • Exhibits anti-tumor activities
  • Inhibits OCI-Ly1 cell line growth with a GI50 of 2.8 μM
  • Inhibits OCI-Ly3 cell line growth with a GI50 of 4.2 μM