Learn More
Medchemexpress LLC Bcl6-in-4 | 2253879-65-9 | 99.7% | 503.04 | 50 MG
SDP
Catalog No. 5000348896
Encompass_Preferred
BCL6-IN-4 is a potent inhibitor of B-cell lymphoma 6 (BCL6) with an IC50 of 97 nM. It demonstrates anti-tumor activities. This product is intended for research use only and is not for medical, clinical, or food use in humans or animals.
  • Potently inhibits B-cell lymphoma 6 (BCL6) with an IC50 of 97 nM.
  • Exhibits anti-tumor properties.
  • Induces BCL6 degradation in human OCI-LY1 cells and shows antiproliferative activity against OCI-LY1 and OCI-LY3 cell lines.
  • Boasts a purity of 99.72%.
  • Primarily for research purposes.
Catalog No. 50-003-48896 Supplier Medchemexpress LLC Supplier No. HY13664050MG
Add to Cart
Add to Cart
BCL6-IN-4 is a potent inhibitor of B-cell lymphoma 6 (BCL6) with an IC50 of 97 nM. It demonstrates anti-tumor activities. This product is intended for research use only and is not for medical, clinical, or food use in humans or animals.
  • Potently inhibits B-cell lymphoma 6 (BCL6) with an IC50 of 97 nM.
  • Exhibits anti-tumor properties.
  • Induces BCL6 degradation in human OCI-LY1 cells and shows antiproliferative activity against OCI-LY1 and OCI-LY3 cell lines.
  • Boasts a purity of 99.72%.
  • Primarily for research purposes.