Learn More
Abnova™ BCL6 Split FISH Probe
Catalog No. 89999065 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 uL
Catalog No. Quantity
89-999-065 100 uL
1 options
Catalog No. 89-999-065 Supplier Abnova™ Supplier No. FS0068
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Quantity 100 uL
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
For Use With (Application) FISH-Ce
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe