Learn More
New England Biolabs, Inc. BclI – 3000 units
SDP
Catalog No. 50812062
Encompass
A restriction endonuclease that recognizes the sequence T^GATC_A.
Catalog No. 50-812-062 Supplier New England Biolabs, Inc. Supplier No. R0160S
A restriction endonuclease that recognizes the sequence T^GATC_A.