Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 598MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 2H12. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a. Positive Control: Jurkat, K562, HL-60, or HeLa Cells. Reed-Sternberg cells in Hodgkin.’s lymphoma. Cellular Staining: Cytoplasmic, cell/nuclear membrane. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 27kDa.Chromosome: 20q11.21. Gene Symbol: BCL2L1. Gene ID: 598. Ensemble: ENSG00000171552. Uniprot: NP_612815.
Catalog No. 50-188-5136