Learn More
ACROBiosystems Recombinant Protein;Human Bcl-x / BCL2L1 Protein, GST Tag;E.coli;1MG;BC1,BCL2L1,Bcl-XL,Bcl2-L-1,BCL-XL/S,BCL2L,BCLX,BCLXL,BCLXS,Bcl-X,bcl-xS
Catalog No. 501659018
Encompass_Preferred
Recombinant Protein;Human Bcl-x / BCL2L1 Protein, GST Tag;1MG;Human Bcl-x, GST Tag (BC1-H513a) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession # AAH19307).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 50.5 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9018 Supplier ACROBiosystems Supplier No. BC1H513A1MG
Recombinant Protein;Human Bcl-x / BCL2L1 Protein, GST Tag;1MG;Human Bcl-x, GST Tag (BC1-H513a) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession # AAH19307).;This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 50.5 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE).