Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Bcl-x Protein, His Tag, Lyophilized, 100 μg

Catalog No. 501659019
Encompass_Preferred

Human Bcl-x, also known as BCL2L1 or Bcl-XL, is a homodimer protein belonging to the Bcl-2 family that acts as a potent inhibitor of cell death by blocking the voltage-dependent anion channel (VDAC) and inhibiting caspase activation. This recombinant protein is expressed in E. coli cells and includes a C-terminal polyhistidine tag. It serves as a regulator of the G2 checkpoint and progression to cytokinesis during mitosis.

  • Purity: >95% as determined by SDS-PAGE
  • Calculated molecular weight: 24.6 kDa
  • Apparent molecular weight: 25–27 kDa under reducing conditions
  • Endotoxin level: <1.0 EU per μg by LAL/rFC method
  • Formulation: Lyophilized from 0.22 μm filtered solution in 20 mM HEPES, 50 mM KCl, pH 7.5 with trehalose

Catalog No. 50-165-9019 Supplier ACROBiosystems Supplier No. BC1H5120100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Bcl-x, also known as BCL2L1 or Bcl-XL, is a homodimer protein belonging to the Bcl-2 family that acts as a potent inhibitor of cell death by blocking the voltage-dependent anion channel (VDAC) and inhibiting caspase activation. This recombinant protein is expressed in E. coli cells and includes a C-terminal polyhistidine tag. It serves as a regulator of the G2 checkpoint and progression to cytokinesis during mitosis.

  • Purity: >95% as determined by SDS-PAGE
  • Calculated molecular weight: 24.6 kDa
  • Apparent molecular weight: 25–27 kDa under reducing conditions
  • Endotoxin level: <1.0 EU per μg by LAL/rFC method
  • Formulation: Lyophilized from 0.22 μm filtered solution in 20 mM HEPES, 50 mM KCl, pH 7.5 with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.