Learn More
ACROBiosystems Recombinant Protein;Human Bcl-x / BCL2L1 Protein;E.coli;1MG;BC1,BCL2L1,Bcl-XL,Bcl2-L-1,BCL-XL/S,BCL2L,BCLX,BCLXL,BCLXS,Bcl-X,bcl-xS
Supplier:  ACROBiosystems BC1H51201MG
Encompass_Preferred
Recombinant Protein;Human Bcl-x / BCL2L1 Protein;1MG;Human Bcl-x, His Tag (BC1-H5120) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession # AAH19307).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 26 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9020