Learn More
ACROBiosystems BCMA 25ug
Catalog No. 502535476
Encompass_Preferred
Biotinylated Cynomolgus / Rhesus macaque BCMA HisAvitag (BCA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # A0A2K5UD97-1). In the region Met 1 - Ala 53 the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.
Catalog No. 50-253-5476 Supplier ACROBiosystems Supplier No. BCAC82E525UG
Biotinylated Cynomolgus / Rhesus macaque BCMA HisAvitag (BCA-C82E5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # A0A2K5UD97-1). In the region Met 1 - Ala 53 the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.