Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc,Avitag™
Catalog No. 501957331
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus / Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 33.9 kDa. The protein migrates as 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque BCMA, Fc,Avitag (Cat. No. BCA-C82F4) with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-195-7331 Supplier ACROBiosystems Supplier No. BCAC82F425UG
Recombinant Protein;25UG;Biotinylated Cynomolgus / Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 33.9 kDa. The protein migrates as 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque BCMA, Fc,Avitag (Cat. No. BCA-C82F4) with a linear range of 0.2-4 ng/mL (QC tested).