Learn More
AdipoGen BCMA hFc hrec
SDP
Catalog No. 502562926
Encompass_Preferred
Protein. The extracellular domain of human BCMA aa 2-54 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds human and mouse BAFF and APRIL. Purity >95% SDS-PAGE. BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. Its downstream signaling is dependent on NF-kappaB and JNK.
Catalog No. 50-256-2926 Supplier AdipoGen Supplier No. AG40B00803050
Protein. The extracellular domain of human BCMA aa 2-54 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds human and mouse BAFF and APRIL. Purity >95% SDS-PAGE. BCMA is a receptor for BAFF and APRIL. It promotes B cell survival and plays a role in the regulation of humoral immunity. Its downstream signaling is dependent on NF-kappaB and JNK.