Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, His Tag
Supplier:  ACROBiosystems BCAC52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque BCMA, His Tag (BCA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.9 kDa. The protein migrates as 16-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, His Tag (Cat. No. BCA-C52H7) with a linear range of 0.078-0.625 μg/mL (QC tested).;Immobilized Cynomolgus / Rhesus BCMA, His Tag (Cat. No. BCA-C52H7) at 2 μg/mL (100 μL/well) can bind Anti-BCMA MAb (Mouse IgG1, the antibody was co developed by SCT and ACRObiosystems) with a linear range of 6.4-160 ng/mL (Routinely tested).
Catalog No. 50-195-7229