Learn More
Enzo Life Sciences BCMA (human) : Fc (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX522026C050
Encompass_Preferred
Alternative name: TNFRSF 17, B cell maturation protein, CD269. MW: ~40kDa (SDS-PAGE). Concentration: 1mg/ml after reconstitution. Endotoxin content: <0.1EU/µg purified protein (LAL test; BioWhittaker). Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human BCMA (aa 2-54) is fused at the C-terminus to the Fc portion of human IgG1. Specificity: Binds human and mouse BAFF and APRIL. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: Q02223. Application: ELISA. Flow Cytometry. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC0987594