Learn More
Enzo Life Sciences BCMA (human) monoclonal antibody (Vicky-1) (PE conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX804151PEC100
Encompass_Preferred
Clone: Vicky-1. Host: Rat. Isotype: IgG1. Immunogen: Recombinant human BCMA extracellular domain (aa 2-54) (Prod. No. ALX-522-026). UniProt: Q02223. Source: Purified from ascites. Species Reactivity: Human. Crossreactivity: Does not cross-react with mouse BCMA. Applications: Flow Cytometry. Recommended dilutions: Flow Cytometry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Do not freeze. Protect from light.
Catalog No. NC0422086