Learn More
Enzo Life Sciences BCMA (human) monoclonal antibody (Vicky-1) (FITC conjugate) (100tests)
SDP
Supplier:  Enzo Life Sciences ALX804151FT100
Encompass_Preferred
Clone: Vicky-1. Isotype: IgG1. Immunogen: Recombinant human BCMA extracellular domain (aa 2-54) (ALX-522-026). Source: Purified from ascites. Species reactivity: Human. Applications: Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9800