Learn More
Enzo Life Sciences BCMA polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905306100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human BCMA. UniProt ID: Q02223. Species reactivity: Human. Applications: ELISA, IF, IHC (PS), WB. Recommended Dilutions: Immunofluorescence (20µg/ml) Immunohistochemistry (paraffin sections, 10µg/ml) Western Blot (2-5µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-6962