Learn More
Sino Biological Recombinant Human BCMA Protein (ECD, His Tag), HPLC-verified
Catalog No. 502114778
Encompass_Preferred
A DNA sequence encoding the human TNFRSF17 (NP_001183.2) (Met1-Ala54) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4778 Supplier Sino Biological Supplier No. 10620H08H50
A DNA sequence encoding the human TNFRSF17 (NP_001183.2) (Met1-Ala54) was expressed with a polyhistidine tag at the C-terminus.