Learn More
Medchemexpress LLC BCMA/TNFRSF17 Trimer 20ug | 20ug
SDP
Catalog No. 5000187814
Encompass_Preferred
B Cell Maturation Antigen (BCMA) also referred to as TNFRSF17 or CD269 is a transmembrane glycoprotein member of the tumor necrosis factor receptor (TNFR) superfamily BCMA is used as a biomarker for Multiple myeloma (MM) BCMA mainly plays an important role in B cells for their proliferation survival and also differentiates them into plasma cells[1] BCMA/TNFRSF17 Trimer Protein Human (Biotinylated HEK293 His-Avi) is a biotinylated recombinant protein with a C-Terminal Avi label and a C-Terminal His label It consists of 54 amino acids (M1-A54) and is produced in HEK293 cells
Catalog No. 50-001-87814 Supplier Medchemexpress LLC Supplier No. HYP7807120UG
B Cell Maturation Antigen (BCMA) also referred to as TNFRSF17 or CD269 is a transmembrane glycoprotein member of the tumor necrosis factor receptor (TNFR) superfamily BCMA is used as a biomarker for Multiple myeloma (MM) BCMA mainly plays an important role in B cells for their proliferation survival and also differentiates them into plasma cells[1] BCMA/TNFRSF17 Trimer Protein Human (Biotinylated HEK293 His-Avi) is a biotinylated recombinant protein with a C-Terminal Avi label and a C-Terminal His label It consists of 54 amino acids (M1-A54) and is produced in HEK293 cells