Learn More
Genscript Corporation BCMA/TNFRSF17 TrimerBiotin
Catalog No. 5000056957
Encompass
B-cell maturation antigen (BCMA or BCM) also known as tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is a protein that in humans is encoded by the TNFRSF17 gene TNFRSF17 is a cell surface receptor of the TNF receptor superfamily which recognizes B-cell activating factor (BAFF)
Catalog No. 50-000-56957 Supplier Genscript Corporation Supplier No. Z03850100
B-cell maturation antigen (BCMA or BCM) also known as tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is a protein that in humans is encoded by the TNFRSF17 gene TNFRSF17 is a cell surface receptor of the TNF receptor superfamily which recognizes B-cell activating factor (BAFF)