Learn More
Genscript Corporation BCMA/TNFRSF17 TrimerBiotin
Supplier:  Genscript Corporation Z03850500
Encompass
B-cell maturation antigen (BCMA or BCM) also known as tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is a protein that in humans is encoded by the TNFRSF17 gene TNFRSF17 is a cell surface receptor of the TNF receptor superfamily which recognizes B-cell activating factor (BAFF)
Catalog No. 50-000-57518