Learn More
ACROBiosystems Recombinant Protein;Mouse BCMA / TNFRSF17 Protein, Fc Tag;HEK293;1MG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Catalog No. 501658182
Encompass_Preferred
Recombinant Protein;Mouse BCMA / TNFRSF17 Protein, Fc Tag;1MG;Mouse BCMA, Fc Tag (BCA-M5258) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/mL (100 μL/well) can bind Mouse BCMA, Fc Tag (Cat. No. BCA-M5258) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-165-8182 Supplier ACROBiosystems Supplier No. BCAM52581MG
Recombinant Protein;Mouse BCMA / TNFRSF17 Protein, Fc Tag;1MG;Mouse BCMA, Fc Tag (BCA-M5258) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/mL (100 μL/well) can bind Mouse BCMA, Fc Tag (Cat. No. BCA-M5258) with a linear range of 2-20 ng/mL (QC tested).