Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse BCMA / TNFRSF17 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Supplier: ACROBiosystems BCAM82F025UG
Recombinant Protein;Biotinylated Mouse BCMA / TNFRSF17 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Mouse BCMA, Fc Tag (BCA-M82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 34.4 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse BCMA, Fc Tag (Cat. No. BCA-M82F0) with a linear range of 2-39 ng/mL (QC tested).