Learn More
BIOCLONE INC BCMAG MONOMER AVIDIN KIT
Catalog No. NC1389122
Encompass_Preferred
BcMag Monomer Avidin Kit Kit components: 2 ml BcMagTM Monomer Avidin Magnetic beads 20 ml 1x Blocking / Elution Buffer 20 ml 1x Regeneration Buffer 15 ml 10x PBS buffer
Catalog No. NC1389122 Supplier BIOCLONE INC Supplier No. MMI101
BcMag Monomer Avidin Kit Kit components: 2 ml BcMagTM Monomer Avidin Magnetic beads 20 ml 1x Blocking / Elution Buffer 20 ml 1x Regeneration Buffer 15 ml 10x PBS buffer