Learn More
BIOCLONE INC BCMAG Monomeric Avidin Beads, superparamagnetic microspheres, coated with high-purity monomeric avidin, 10 mg/ml concentration, ~2.5 μm diameter
Catalog No. NC1175159
Encompass_Preferred
BCMAG Monomeric Avidin Beads, superparamagnetic microspheres, coated with high-purity monomeric avidin, 10 mg/ml concentration, ~2.5 μm diameter
Catalog No. NC1175159 Supplier BIOCLONE INC Supplier No. MMI102
BCMAG Monomeric Avidin Beads, superparamagnetic microspheres, coated with high-purity monomeric avidin, 10 mg/ml concentration, ~2.5 μm diameter